Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mff | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017226.1 | - | 5 - 6 | 755 | 111 | |
AK139649.1 | - | 5 - 6 | 731 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |