Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026341.2 | NP_080617.2 | 5 - 6 | 577 | 73 | |
GenBank mRNA | AK014204.1 | - | 6 - 7 | 800 | 73 | |
AK017031.1 | - | 5 - 6 | 546 | 73 | ||
AK161922.1 | - | 5 - 6 | 639 | 73 | ||
BC021366.1 | - | 7 - 8 | 843 | 73 | ||
BC037091.1 | - | 4 - 5 | 500 | 73 | ||
BC040807.1 | - | 5 - 6 | 524 | 73 | ||
BC049948.1 | - | 5 - 6 | 531 | 3666660 | 73 | |
CA983915.1 | - | 3 - 4 | 313 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |