Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700018829.mm.1 |
NM_001198833.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018829.mm.1 |
NM_001198833.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018829.mm.1 |
TC1700001976.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700018834.mm.1 |
NM_001198831.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018834.mm.1 |
NM_001198831.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018834.mm.1 |
TC1700001976.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700018832.mm.1 |
TC1700001976.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700018825.mm.1 |
NM_001198831.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018825.mm.1 |
NM_001198831.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018825.mm.1 |
NM_001198833.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018825.mm.1 |
NM_001198833.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018825.mm.1 |
NM_007584.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018825.mm.1 |
NM_007584.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018825.mm.1 |
NM_172962.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018825.mm.1 |
NM_172962.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018825.mm.1 |
TC1700001976.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700018826.mm.1 |
NM_001198831.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018826.mm.1 |
NM_001198831.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018826.mm.1 |
NM_001198833.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018826.mm.1 |
NM_001198833.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018826.mm.1 |
NM_007584.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018826.mm.1 |
NM_007584.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018826.mm.1 |
NM_172962.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018826.mm.1 |
NM_172962.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018826.mm.1 |
TC1700001976.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700018830.mm.1 |
NM_001198833.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018830.mm.1 |
NM_001198833.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700018830.mm.1 |
TC1700001976.mm.1 |
MTA TC |