Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178281.1 | NP_840065.1 | 1 - 2 | 98 | 61 | |
GenBank mRNA | AK053507.1 | - | 1 - 2 | 50 | 61 | |
AK145861.1 | - | 1 - 2 | 49 | 61 | ||
AK220298.1 | - | 1 - 2 | 34 | 61 | ||
BY312131.1 | - | 1 - 2 | 99 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus 1700 Inventoried Probe spans exons |