Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dazl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010021.5 | NP_034151.3 | 7 - 8 | 785 | 126 | |
GenBank mRNA | AK014854.1 | - | 7 - 8 | 765 | 126 | |
AK033183.1 | - | 7 - 8 | 765 | 126 | ||
AK160855.1 | - | 7 - 8 | 644 | 126 | ||
BC099940.1 | - | 7 - 8 | 745 | 30048233 | 126 | |
U38690.1 | - | 7 - 8 | 900 | 126 | ||
U46694.1 | - | 7 - 8 | 681 | 126 | ||
X95724.1 | - | 7 - 8 | 674 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |