Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfeb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161722.1 | NP_001155194.1 | 7 - 8 | 1108 | 72 | |
NM_001161723.1 | NP_001155195.1 | 7 - 8 | 1102 | 72 | ||
NM_011549.3 | NP_035679.3 | 6 - 7 | 1049 | 72 | ||
GenBank mRNA | AF079095.1 | - | 6 - 7 | 1030 | 72 | |
AK146016.1 | - | 6 - 7 | 1074 | 72 | ||
AK154971.1 | - | 7 - 8 | 1106 | 72 | ||
BC064789.1 | - | 6 - 7 | 1049 | 30429545 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |