Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110832.1 | NP_001104302.1 | 5 - 6 | 704 | 82 | |
NM_010913.2 | NP_035043.1 | 4 - 5 | 617 | 82 | ||
GenBank mRNA | AK004729.1 | - | 5 - 6 | 650 | 82 | |
AK143723.1 | - | 4 - 5 | 579 | 82 | ||
AK156487.1 | - | 5 - 6 | 534 | 82 | ||
BC029695.1 | - | 4 - 5 | 505 | 5374597 | 82 | |
BC057099.1 | - | 5 - 6 | 606 | 5716111 | 82 | |
M86214.1 | - | 3 - 4 | 354 | 82 | ||
M86215.1 | - | 4 - 5 | 438 | 82 | ||
X55315.1 | - | 4 - 5 | 527 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |