Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142744.1 | NP_001136216.1 | 12 - 13 | 1127 | 76 | |
GenBank mRNA | AK011750.2 | - | 12 - 13 | 1103 | 76 | |
AK011838.1 | - | 12 - 13 | 1118 | 76 | ||
AK028297.1 | - | 11 - 12 | 1088 | 76 | ||
BC028847.1 | - | 12 - 13 | 1106 | 4007862 | 76 | |
BC030669.1 | - | 11 - 12 | 1036 | 1246856 | 76 | |
BQ748472.1 | - | 3 - 4 | 163 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |