Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mocs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020042.2 | NP_064426.2 | 5 - 6 | 724 | 91 | |
NM_028464.1 | NP_082740.1 | 5 - 6 | 724 | 91 | ||
GenBank mRNA | AK157231.1 | - | 5 - 6 | 680 | 91 | |
BC034066.1 | - | 1 - 2 | 73 | 91 | ||
BC049914.1 | - | 3 - 4 | 352 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |