Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mocs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020042.2 | NP_064426.2 | 8 - 9 | 1054 | 70 | |
NM_028464.1 | NP_082740.1 | 8 - 9 | 1054 | 70 | ||
GenBank mRNA | AF214016.1 | - | 2 - 3 | 229 | 70 | |
AK157231.1 | - | 8 - 9 | 1010 | 70 | ||
BC034066.1 | - | 4 - 5 | 403 | 70 | ||
BC049914.1 | - | 6 - 7 | 682 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |