Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tjap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252473.1 | NP_001239402.1 | 1 - 2 | 67 | 83 | |
NM_001252474.1 | NP_001239403.1 | 1 - 2 | 67 | 83 | ||
NM_028751.3 | NP_083027.1 | 1 - 2 | 67 | 83 | ||
GenBank mRNA | AK002882.2 | - | 1 - 2 | 54 | 83 | |
AK164925.1 | - | 1 - 2 | 52 | 83 | ||
AK169889.1 | - | 1 - 2 | 42 | 83 | ||
BC023316.1 | - | 1 - 2 | 53 | 3993257 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |