Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tjap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252473.1 | NP_001239402.1 | 6 - 7 | 577 | 63 | |
NM_001252474.1 | NP_001239403.1 | 6 - 7 | 574 | 63 | ||
NM_001252475.1 | NP_001239404.1 | 5 - 6 | 480 | 63 | ||
NM_028751.3 | NP_083027.1 | 7 - 8 | 707 | 63 | ||
GenBank mRNA | AB712252.1 | - | 3 - 4 | 294 | 63 | |
AK002882.2 | - | 7 - 8 | 694 | 63 | ||
AK164925.1 | - | 6 - 7 | 559 | 63 | ||
AK169889.1 | - | 6 - 7 | 553 | 63 | ||
BC023316.1 | - | 7 - 8 | 693 | 3993257 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |