Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010833.2 | NP_001010833.2 | 10 - 11 | 4512 | 63 | |
GenBank mRNA | AK129074.1 | - | 4 - 5 | 2248 | 63 | |
BC085140.1 | - | 5 - 6 | 2381 | 63 | ||
BC094363.1 | - | 6 - 7 | 3215 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |