Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253885.1 | NP_001240814.1 | 6 - 7 | 1476 | 83 | |
NM_001253886.1 | NP_001240815.1 | 6 - 7 | 1476 | 83 | ||
NM_001253887.1 | NP_001240816.1 | 6 - 7 | 476 | 83 | ||
NM_009685.3 | NP_033815.1 | 6 - 7 | 1290 | 83 | ||
GenBank mRNA | AK030748.1 | - | 6 - 7 | 1173 | 83 | |
AK075730.1 | - | 6 - 7 | 456 | 83 | ||
AK083830.1 | - | 6 - 7 | 1206 | 83 | ||
AK144340.1 | - | 6 - 7 | 1477 | 83 | ||
AK164140.1 | - | 6 - 7 | 1201 | 83 | ||
BC036956.1 | - | 5 - 6 | 1114 | 83 | ||
BC048395.1 | - | 4 - 5 | 622 | 6392016 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |