Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082552.2 | NP_001076021.1 | 2 - 3 | 527 | 79 | |
NM_009277.4 | NP_033303.3 | 3 - 4 | 596 | 79 | ||
GenBank mRNA | AK141294.1 | - | 3 - 4 | 575 | 79 | |
AK150484.1 | - | 2 - 3 | 491 | 79 | ||
AK152619.1 | - | 2 - 3 | 491 | 79 | ||
AK171489.1 | - | 3 - 4 | 560 | 79 | ||
BC010580.1 | - | 3 - 4 | 536 | 3584654 | 79 | |
CT010336.1 | - | 1 - 2 | 421 | 79 | ||
L27990.1 | - | 3 - 4 | 561 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |