Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082552.2 | NP_001076021.1 | 3 - 4 | 624 | 67 | |
NM_009277.4 | NP_033303.3 | 4 - 5 | 693 | 67 | ||
GenBank mRNA | AK141294.1 | - | 4 - 5 | 672 | 67 | |
AK150484.1 | - | 3 - 4 | 588 | 67 | ||
AK152619.1 | - | 3 - 4 | 588 | 67 | ||
AK171489.1 | - | 4 - 5 | 657 | 67 | ||
BC010580.1 | - | 4 - 5 | 633 | 3584654 | 67 | |
CT010336.1 | - | 2 - 3 | 518 | 67 | ||
L27990.1 | - | 4 - 5 | 658 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |