Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153527.2 | NP_705755.2 | 2 - 3 | 224 | 77 | |
GenBank mRNA | AF419292.2 | - | 2 - 3 | 369 | 77 | |
AK005984.1 | - | 2 - 3 | 241 | 77 | ||
AK139680.1 | - | 196 | 77 | |||
BC048501.1 | - | 2 - 3 | 224 | 6705915 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |