Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400011708.mm.1 |
NM_001007268.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400011708.mm.1 |
NM_001007268.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400011708.mm.1 |
NM_013849.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400011708.mm.1 |
NM_013849.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400011708.mm.1 |
TC1400001558.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400011707.mm.1 |
NM_001007268.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400011707.mm.1 |
NM_001007268.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400011707.mm.1 |
NM_013849.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400011707.mm.1 |
NM_013849.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400011707.mm.1 |
TC1400001558.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400011710.mm.1 |
NM_001007268.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400011710.mm.1 |
NM_001007268.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400011710.mm.1 |
NM_013849.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400011710.mm.1 |
NM_013849.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400011710.mm.1 |
TC1400001558.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400011717.mm.1 |
NM_013849.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400011717.mm.1 |
NM_013849.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400011717.mm.1 |
TC1400001558.mm.1 |
MTA TC |