Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001326.1 | NP_001001326.1 | 1 - 2 | 91 | 61 | |
NM_029811.2 | NP_084087.2 | 1 - 2 | 91 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |