Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040131.2 | NP_001035221.1 | 9 - 10 | 1146 | 68 | |
NM_013507.3 | NP_038535.2 | 9 - 10 | 1146 | 68 | ||
GenBank mRNA | AK141512.1 | - | 2 - 3 | 216 | 68 | |
AK144309.1 | - | 9 - 10 | 1120 | 68 | ||
AK150745.1 | - | 9 - 10 | 1123 | 68 | ||
AK151637.1 | - | 9 - 10 | 1123 | 68 | ||
BC039274.1 | - | 8 - 9 | 824 | 68 | ||
BC040391.1 | - | 8 - 9 | 835 | 4018962 | 68 | |
BC043034.1 | - | 8 - 9 | 826 | 5696921 | 68 | |
BC056387.1 | - | 9 - 10 | 1110 | 6854755 | 68 | |
BC057673.1 | - | 10 - 11 | 998 | 4020759 | 68 | |
BC064810.1 | - | 8 - 9 | 879 | 6825556 | 68 | |
BC092521.1 | - | 9 - 10 | 1101 | 30649919 | 68 | |
BC127064.1 | - | 9 - 10 | 1146 | 40109978 | 68 | |
BU610672.1 | - | 1 - 2 | 128 | 68 | ||
U63323.1 | - | 9 - 10 | 1098 | 68 | ||
U76112.1 | - | 9 - 10 | 1120 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |