Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040131.2 | NP_001035221.1 | 10 - 11 | 1228 | 59 | |
NM_013507.3 | NP_038535.2 | 10 - 11 | 1228 | 59 | ||
GenBank mRNA | AK141512.1 | - | 3 - 4 | 298 | 59 | |
AK144309.1 | - | 10 - 11 | 1202 | 59 | ||
AK150745.1 | - | 10 - 11 | 1205 | 59 | ||
AK151637.1 | - | 10 - 11 | 1205 | 59 | ||
BC039274.1 | - | 9 - 10 | 906 | 59 | ||
BC040391.1 | - | 9 - 10 | 917 | 4018962 | 59 | |
BC043034.1 | - | 9 - 10 | 908 | 5696921 | 59 | |
BC056387.1 | - | 10 - 11 | 1192 | 6854755 | 59 | |
BC057673.1 | - | 11 - 12 | 1080 | 4020759 | 59 | |
BC064810.1 | - | 9 - 10 | 961 | 6825556 | 59 | |
BC092521.1 | - | 10 - 11 | 1183 | 30649919 | 59 | |
BC127064.1 | - | 10 - 11 | 1228 | 40109978 | 59 | |
BU610672.1 | - | 2 - 3 | 210 | 59 | ||
U63323.1 | - | 10 - 11 | 1180 | 59 | ||
U76112.1 | - | 10 - 11 | 1202 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |