Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173767.3 | NP_776128.2 | 5 - 6 | 861 | 106 | |
GenBank mRNA | AK014452.1 | - | 4 - 5 | 622 | 106 | |
AK050143.1 | - | 2 - 3 | 356 | 106 | ||
AK187129.1 | - | 1 - 2 | 106 | |||
BC026405.1 | - | 5 - 6 | 748 | 4211657 | 106 | |
BC089586.1 | - | 2 - 3 | 365 | 106 | ||
BC139196.1 | - | 2 - 3 | 249 | 8862218 | 106 | |
BC139197.1 | - | 2 - 3 | 249 | 8862219 | 106 | |
DQ205645.1 | - | 4 - 5 | 574 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |