Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033954.3 | NP_001029126.1 | 3 - 4 | 382 | 71 | |
NM_001289444.1 | NP_001276373.1 | 3 - 4 | 406 | 71 | ||
GenBank mRNA | AF330212.1 | - | 3 - 4 | 320 | 71 | |
BC028771.1 | - | 3 - 4 | 315 | 1331673 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |