Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025559.3 | NP_001020730.1 | 14 - 15 | 1985 | 72 | |
NM_001025560.2 | NP_001020731.1 | 14 - 15 | 2099 | 72 | ||
NM_001277326.1 | NP_001264255.1 | 14 - 15 | 2080 | 72 | ||
NM_001277327.1 | NP_001264256.1 | 13 - 14 | 2140 | 72 | ||
NM_001277328.1 | NP_001264257.1 | 12 - 13 | 1845 | 72 | ||
NM_011445.4 | NP_035575.1 | 15 - 16 | 2133 | 72 | ||
GenBank mRNA | AJ010605.1 | - | 3 - 4 | 412 | 72 | |
AK029696.1 | - | 12 - 13 | 1773 | 72 | ||
AK030713.1 | - | 14 - 15 | 2100 | 72 | ||
AK132409.1 | - | 14 - 15 | 2046 | 72 | ||
BC067407.1 | - | 13 - 14 | 2137 | 30094413 | 72 | |
D61689.1 | - | 12 - 13 | 1841 | 72 | ||
U32614.1 | - | 15 - 16 | 2137 | 72 | ||
X65659.1 | - | 1 - 2 | 74 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |