Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025559.3 | NP_001020730.1 | 4 - 5 | 582 | 143 | |
NM_001025560.2 | NP_001020731.1 | 4 - 5 | 702 | 143 | ||
NM_001277326.1 | NP_001264255.1 | 3 - 4 | 557 | 143 | ||
NM_001277327.1 | NP_001264256.1 | 3 - 4 | 737 | 143 | ||
NM_001277328.1 | NP_001264257.1 | 2 - 3 | 445 | 143 | ||
NM_011445.4 | NP_035575.1 | 4 - 5 | 610 | 143 | ||
GenBank mRNA | AK029696.1 | - | 2 - 3 | 372 | 143 | |
AK030713.1 | - | 4 - 5 | 703 | 143 | ||
AK048143.1 | - | 4 - 5 | 583 | 143 | ||
AK084290.1 | - | 4 - 5 | 741 | 143 | ||
AK132409.1 | - | 3 - 4 | 523 | 143 | ||
BC067407.1 | - | 3 - 4 | 734 | 30094413 | 143 | |
D61689.1 | - | 2 - 3 | 441 | 143 | ||
U32614.1 | - | 4 - 5 | 614 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |