Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016870.3 | NP_058566.3 | 3 - 4 | 413 | 106 | |
NM_212441.2 | NP_997606.2 | 2 - 3 | 308 | 106 | ||
NM_212442.2 | NP_997607.2 | 2 - 3 | 211 | 106 | ||
GenBank mRNA | AF068246.1 | - | 2 - 3 | 199 | 106 | |
AK041060.1 | - | 2 - 3 | 183 | 106 | ||
AK077247.1 | - | 2 - 3 | 270 | 106 | ||
AK143946.1 | - | 211 | 106 | |||
AK165516.1 | - | 2 - 3 | 185 | 106 | ||
AY064696.1 | - | 2 - 3 | 284 | 106 | ||
BB846489.1 | - | 2 - 3 | 212 | 106 | ||
BC015248.1 | - | 3 - 4 | 373 | 4239288 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |