Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008855.2 | NP_032881.1 | 9 - 10 | 1285 | 88 | |
GenBank mRNA | AK051862.1 | - | 9 - 10 | 1278 | 88 | |
BC127083.1 | - | 9 - 10 | 1104 | 40111744 | 88 | |
X53532.1 | - | 9 - 10 | 1113 | 88 | ||
X59274.1 | - | 9 - 10 | 1101 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |