Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145589.2 | NP_663564.2 | 6 - 7 | 1093 | 85 | |
GenBank mRNA | AK133884.1 | - | 6 - 7 | 1057 | 85 | |
AK139559.1 | - | 7 - 8 | 868 | 85 | ||
AK145816.1 | - | 7 - 8 | 873 | 85 | ||
AK151203.1 | - | 7 - 8 | 925 | 85 | ||
BC055033.1 | - | 6 - 7 | 1093 | 6310272 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |