Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290740.1 | NP_001277669.1 | 16 - 17 | 2002 | 124 | |
NM_030560.5 | NP_085037.2 | 17 - 18 | 2060 | 124 | ||
NM_172667.2 | NP_766255.1 | 16 - 17 | 1999 | 124 | ||
GenBank mRNA | AK045589.1 | - | 16 - 17 | 1831 | 124 | |
AK077961.1 | - | 17 - 18 | 1888 | 124 | ||
AK129404.1 | - | 3144 | 124 | |||
AK132001.1 | - | 16 - 17 | 1943 | 124 | ||
AK149528.1 | - | 16 - 17 | 1826 | 124 | ||
BC003273.1 | - | 9 - 10 | 926 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |