Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700037730.mm.1 |
NM_001190385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037730.mm.1 |
NM_001190385.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037730.mm.1 |
NM_026769.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037730.mm.1 |
NM_026769.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037730.mm.1 |
TC0700004492.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700037729.mm.1 |
NM_001190385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037729.mm.1 |
NM_001190385.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037729.mm.1 |
NM_026769.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037729.mm.1 |
NM_026769.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037729.mm.1 |
TC0700004492.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700037722.mm.1 |
NM_001190385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037722.mm.1 |
NM_001190385.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037722.mm.1 |
NM_001190386.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037722.mm.1 |
NM_001190386.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037722.mm.1 |
NM_026769.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037722.mm.1 |
NM_026769.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700037722.mm.1 |
TC0700004492.mm.1 |
MTA TC |