Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021282.2 | NP_067257.1 | 2 - 3 | 348 | 105 | |
GenBank mRNA | BC013451.1 | - | 2 - 3 | 348 | 4223797 | 105 |
BC042693.1 | - | 2 - 3 | 327 | 105 | ||
DQ838976.1 | - | 2 - 3 | 335 | 105 | ||
L11650.1 | - | 2 - 3 | 441 | 105 | ||
X62595.1 | - | 2 - 3 | 333 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |