Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163689.1 | NP_001157161.1 | 6 - 7 | 1026 | 60 | |
NR_028142.1 | 7 - 8 | 1108 | 60 | |||
GenBank mRNA | AK031609.1 | - | 6 - 7 | 984 | 60 | |
AK076464.1 | - | 6 - 7 | 810 | 60 | ||
AK085926.1 | - | 6 - 7 | 1011 | 60 | ||
AK150184.1 | - | 6 - 7 | 1008 | 60 | ||
AY510273.1 | - | 6 - 7 | 1007 | 60 | ||
AY894805.1 | - | 6 - 7 | 927 | 60 | ||
BC019188.1 | - | 7 - 8 | 1090 | 5027544 | 60 | |
BC034190.1 | - | 5 - 6 | 527 | 60 | ||
BC044737.1 | - | 7 - 8 | 1072 | 60 | ||
BC044781.2 | - | 7 - 8 | 1072 | 5151911 | 60 | |
BC054825.1 | - | 7 - 8 | 988 | 60 | ||
BC064747.1 | - | 6 - 7 | 998 | 30024535 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |