Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289845.1 | NP_001276774.1 | 2 - 3 | 279 | 103 | |
GenBank mRNA | BC055302.1 | - | 2 - 3 | 242 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |