Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700015540.mm.1 |
TC0700001818.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015540.mm.1 |
NM_026884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015540.mm.1 |
NM_026884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015537.mm.1 |
TC0700001818.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015537.mm.1 |
NM_026884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015537.mm.1 |
NM_026884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015535.mm.1 |
TC0700001818.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015541.mm.1 |
TC0700001818.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015541.mm.1 |
NM_026884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015541.mm.1 |
NM_026884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015539.mm.1 |
TC0700001818.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015539.mm.1 |
NM_026884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015539.mm.1 |
NM_026884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015543.mm.1 |
TC0700001818.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015543.mm.1 |
NM_026884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015543.mm.1 |
NM_026884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015547.mm.1 |
TC0700001818.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015547.mm.1 |
NM_001146347.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015547.mm.1 |
NM_001146347.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015547.mm.1 |
NM_026884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015547.mm.1 |
NM_026884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015547.mm.1 |
NM_029978.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015547.mm.1 |
NM_029978.1 |
Refseq |