Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam53b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK147725.1 | - | 1 - 2 | 254 | 77 | |
AK194861.1 | - | 1 - 2 | 239 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |