Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2064841 |
NM_001081268.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2064841 |
AK220244.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2064841 |
NM_172749.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2064841 |
AK078520.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2064841 |
AK129112.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
730898 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
633981 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
671593 |
AK089249.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
671593 |
AK089470.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
671593 |
NM_010705.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
24939 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P161297 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2064841 |
NM_001081268.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2064841 |
AK220244.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2064841 |
NM_172749.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2064841 |
AK078520.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2064841 |
AK129112.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P501167 |
NM_001081268.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P501167 |
AK220244.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P501167 |
BC039632.1 |
mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507863 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507907 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507874 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507952 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507824 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507883 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507825 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507870 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507994 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507917 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507829 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507876 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507947 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507916 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507908 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507964 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507830 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507836 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507878 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507898 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507827 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507945 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507845 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507867 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507926 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507816 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507937 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507929 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507949 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507837 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507865 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507981 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507849 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507818 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507869 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507923 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507921 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507880 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507976 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507832 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507932 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507986 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507900 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507840 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507973 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507905 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507963 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507992 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507927 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507872 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507822 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507939 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507842 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507858 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507919 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507861 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507834 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507853 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507944 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507943 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507910 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507909 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507983 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507847 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507881 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507879 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25507912 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06290960 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507929 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507949 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507951 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06290980 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507928 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507926 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507953 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507916 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507905 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507940 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507917 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507923 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06290985 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06284997 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507935 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507902 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507908 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06290953 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507907 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06284996 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507909 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507919 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507945 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06291010 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507950 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507915 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06290998 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06095853 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507925 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507937 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507898 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507910 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507952 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507948 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06290993 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507939 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25507954 |
- |
- |