Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039534.1 | NP_001034623.1 | 4 - 5 | 818 | 120 | |
GenBank mRNA | AI173292.1 | - | 2 - 3 | 314 | 120 | |
BC030313.1 | - | 2 - 3 | 1310 | 120 | ||
BC110381.1 | - | 4 - 5 | 794 | 5149569 | 120 | |
BC115503.2 | - | 4 - 5 | 819 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |