Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028724.4 | NP_083000.5 | 7 - 8 | 788 | 66 | |
GenBank mRNA | AK151332.1 | - | 4 - 5 | 552 | 66 | |
AK152893.1 | - | 5 - 6 | 596 | 66 | ||
BC040390.1 | - | 7 - 8 | 782 | 3156682 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |