Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200040746.mm.1 |
NM_001199188.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040746.mm.1 |
NM_001199188.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040746.mm.1 |
TC0200004756.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040743.mm.1 |
NM_001199188.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040743.mm.1 |
NM_001199188.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040743.mm.1 |
NM_024225.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040743.mm.1 |
NM_024225.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040743.mm.1 |
TC0200004756.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040745.mm.1 |
NM_001199188.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040745.mm.1 |
NM_001199188.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040745.mm.1 |
TC0200004756.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200040747.mm.1 |
NM_001199188.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040747.mm.1 |
NM_001199188.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200040747.mm.1 |
TC0200004756.mm.1 |
MTA TC |