Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178755.3 | NP_848870.2 | 16 - 17 | 2489 | 81 | |
GenBank mRNA | AK029744.1 | - | 16 - 17 | 2522 | 81 | |
AK029932.1 | - | 16 - 17 | 2490 | 81 | ||
AK030138.1 | - | 16 - 17 | 2477 | 81 | ||
AK030182.1 | - | 16 - 17 | 2536 | 81 | ||
AK041286.1 | - | 8 - 9 | 898 | 81 | ||
DQ867026.1 | - | 16 - 17 | 2522 | 81 | ||
DQ867027.1 | - | 16 - 17 | 2522 | 81 | ||
DQ867028.1 | - | 16 - 17 | 2522 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |