Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178755.3 | NP_848870.2 | 2 - 3 | 115 | 91 | |
GenBank mRNA | AK029744.1 | - | 2 - 3 | 148 | 91 | |
AK029932.1 | - | 2 - 3 | 116 | 91 | ||
DQ867026.1 | - | 2 - 3 | 148 | 91 | ||
DQ867027.1 | - | 2 - 3 | 148 | 91 | ||
DQ867028.1 | - | 2 - 3 | 148 | 91 | ||
DQ867029.1 | - | 2 - 3 | 148 | 91 | ||
DQ867030.1 | - | 2 - 3 | 148 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |