Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK078278.1 | - | 218 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |