Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146211.1 | NP_001139683.1 | 1 - 2 | 69 | 141 | |
NM_001146212.1 | NP_001139684.1 | 1 - 2 | 69 | 144 | ||
GenBank mRNA | BC038917.1 | - | 1 - 2 | 59 | 141 | |
DV046794.1 | - | 1 - 2 | 83 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |