Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025599.3 | NP_001020770.2 | 6 - 7 | 1049 | 72 | |
NM_001286726.1 | NP_001273655.1 | 6 - 7 | 1068 | 72 | ||
NM_001286727.1 | NP_001273656.1 | 6 - 7 | 1017 | 72 | ||
GenBank mRNA | AK042252.1 | - | 6 - 7 | 1032 | 72 | |
AK148247.1 | - | 6 - 7 | 72 | |||
BC037110.1 | - | 6 - 7 | 1031 | 5369645 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |