Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172497.3 | NP_766085.2 | 4 - 5 | 1274 | 81 | |
GenBank mRNA | AK029550.1 | - | 4 - 5 | 1275 | 81 | |
BC132103.1 | - | 4 - 5 | 1265 | 40130380 | 81 | |
BC137880.1 | - | 4 - 5 | 1265 | 8860899 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |