Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175649.5 | NP_783580.1 | 3 - 4 | 260 | 106 | |
GenBank mRNA | AK142511.1 | - | 321 | 106 | ||
AK154666.1 | - | 3 - 4 | 371 | 106 | ||
AY165628.1 | - | 3 - 4 | 260 | 106 | ||
BC111108.1 | - | 3 - 4 | 248 | 106 | ||
BC138819.1 | - | 3 - 4 | 309 | 8861841 | 106 | |
BC138820.1 | - | 3 - 4 | 310 | 8861842 | 106 | |
BC152357.1 | - | 3 - 4 | 256 | 106 | ||
BC152358.1 | - | 3 - 4 | 260 | 40055748 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |