Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048176.1 | NP_001041641.1 | 4 - 5 | 723 | 66 | |
GenBank mRNA | BC046474.1 | - | 4 - 5 | 829 | 66 | |
CJ130836.1 | - | 3 - 4 | 302 | 66 | ||
HQ426680.1 | - | 4 - 5 | 723 | 66 | ||
HQ426684.1 | - | 3 - 4 | 591 | 66 | ||
HQ426685.1 | - | 4 - 5 | 723 | 66 | ||
HQ426687.1 | - | 4 - 5 | 854 | 66 | ||
HQ426688.1 | - | 4 - 5 | 723 | 66 | ||
HQ426689.1 | - | 3 - 4 | 591 | 66 | ||
HQ426690.1 | - | 3 - 4 | 591 | 66 | ||
HQ426691.1 | - | 522 | 66 | |||
HQ426692.1 | - | 653 | 66 | |||
HQ426693.1 | - | 563 | 66 | |||
HQ426694.1 | - | 476 | 66 | |||
HQ426695.1 | - | 695 | 66 | |||
HQ426696.1 | - | 695 | 66 | |||
HQ426697.1 | - | 695 | 66 | |||
HQ426700.1 | - | 826 | 66 | |||
HQ426701.1 | - | 695 | 66 | |||
HQ426702.1 | - | 3 - 4 | 305 | 66 | ||
HQ426704.1 | - | 3 - 4 | 305 | 66 | ||
HQ426706.1 | - | 2 - 3 | 249 | 66 | ||
HQ426707.1 | - | 2 - 3 | 249 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |