Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048176.1 | NP_001041641.1 | 6 - 7 | 948 | 100 | |
GenBank mRNA | BC046474.1 | - | 6 - 7 | 1054 | 100 | |
DT912627.1 | - | 2 - 3 | 144 | 100 | ||
HQ426680.1 | - | 6 - 7 | 948 | 100 | ||
HQ426681.1 | - | 4 - 5 | 741 | 100 | ||
HQ426682.1 | - | 3 - 4 | 609 | 100 | ||
HQ426683.1 | - | 2 - 3 | 366 | 100 | ||
HQ426684.1 | - | 5 - 6 | 816 | 100 | ||
HQ426685.1 | - | 6 - 7 | 948 | 100 | ||
HQ426686.1 | - | 5 - 6 | 805 | 100 | ||
HQ426687.1 | - | 6 - 7 | 1079 | 100 | ||
HQ426688.1 | - | 6 - 7 | 1095 | 100 | ||
HQ426689.1 | - | 5 - 6 | 963 | 100 | ||
HQ426690.1 | - | 6 - 7 | 1165 | 100 | ||
HQ426691.1 | - | 747 | 100 | |||
HQ426692.1 | - | 878 | 100 | |||
HQ426693.1 | - | 788 | 100 | |||
HQ426694.1 | - | 701 | 100 | |||
HQ426695.1 | - | 920 | 100 | |||
HQ426696.1 | - | 920 | 100 | |||
HQ426697.1 | - | 920 | 100 | |||
HQ426698.1 | - | 713 | 100 | |||
HQ426699.1 | - | 777 | 100 | |||
HQ426700.1 | - | 1051 | 100 | |||
HQ426701.1 | - | 1067 | 100 | |||
HQ426702.1 | - | 5 - 6 | 530 | 100 | ||
HQ426703.1 | - | 3 - 4 | 323 | 100 | ||
HQ426704.1 | - | 5 - 6 | 530 | 100 | ||
HQ426705.1 | - | 4 - 5 | 387 | 100 | ||
HQ426706.1 | - | 4 - 5 | 474 | 100 | ||
HQ426707.1 | - | 4 - 5 | 621 | 100 | ||
HQ426709.1 | - | 2 - 3 | 222 | 100 | ||
HQ426710.1 | - | 2 - 3 | 222 | 100 | ||
HQ426711.1 | - | 2 - 3 | 369 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |