Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026115.4 | NP_080391.2 | 3 - 4 | 219 | 131 | |
GenBank mRNA | AK010715.1 | - | 3 - 4 | 228 | 131 | |
AK014330.1 | - | 3 - 4 | 231 | 131 | ||
AK018270.1 | - | 3 - 4 | 214 | 131 | ||
AK041700.1 | - | 3 - 4 | 223 | 131 | ||
AK137648.1 | - | 3 - 4 | 237 | 131 | ||
AK169315.1 | - | 3 - 4 | 222 | 131 | ||
AK182724.1 | - | 2 - 3 | 176 | 131 | ||
BC055460.1 | - | 2 - 3 | 186 | 4457298 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |