Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172436.3 | NP_766024.1 | 13 - 14 | 1333 | 66 | |
GenBank mRNA | AK019150.1 | - | 13 - 14 | 1336 | 66 | |
AK030595.1 | - | 13 - 14 | 1363 | 66 | ||
AK083156.1 | - | 13 - 14 | 1334 | 66 | ||
AK144958.1 | - | 13 - 14 | 1367 | 66 | ||
AK147948.1 | - | 13 - 14 | 1349 | 66 | ||
BC013845.1 | - | 13 - 14 | 1312 | 66 | ||
BC024952.1 | - | 13 - 14 | 1526 | 66 | ||
BC060505.1 | - | 13 - 14 | 1320 | 6311836 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |